(±)-2-Ethyl-1-hexanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
2-Ethylhexan-1-ol
M-Clarity™ quality level = MQ200
8.00990.0100
8.00990.1000
8.00990.2500
EM8.00990.0100CS
1184
USD
EM8.00990.0100
EM8.00990.1000
EM8.00990.2500
(±)-2-Ethyl-1-hexanol for synthesis, Sigma-Aldrich®
(±)-2-Ethyl-1-hexanol
Formula:
CH₃(CH₂)₃CH(C₂H₅)CH₂OH MW: 130.23 g/mol Boiling Pt: 183 Melting Pt: -76 Flash Pt: 81 |
MDL Number:
MFCD00004746 CAS Number: 104-76-7 Merck Index: 13,03842 |
|
Specification Test Results
Solubility | 1.1 g/l (20 °C) |
Melting Point | -76 °C |
Molar Mass | 130.23 g/mol |
Boiling Point | 184 °C (1013 hPa) |
Vapor Pressure | 0.13 hPa (20 °C) |
Flash Point | 75 °C DIN 51758 |
Refractive Index | 1.4317 (20 °C, 589 nm) |
Explosion Limit | 1.1 - 12.7 %(V) |
Density | 0.832 g/cm3 (20 °C) |
pH | 7 (1 g/l, H2O, 20 °C) |
Ignition Point | 270 - 330 °C DIN 51794 |
Synonyms | Isocotyl alcohol, Isooctanol |
Hill Formula | C8H18O |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...