3-Pentanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Diethyl ketone

M-Clarity™ quality level = MQ200

8.03605.0100 8.03605.1000
EM8.03605.0100CS 1037 USD
EM8.03605.0100 EM8.03605.1000
3-Pentanone for synthesis, Sigma-Aldrich®
3-Pentanone
Formula: C₅H₁₀O
MW: 86.13 g/mol
MDL Number: MFCD00009320
CAS Number: 96-22-0
UN: 1156
ADR: 3,II

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point -40 °C
Molar Mass 86.13 g/mol
Boiling Point 101 - 103 °C (1013 mbar)
Vapor Pressure 37.6 hPa (20 °C)
Flash Point 7 °C
Refractive Index 1.3923 (20 °C, 589 nm)
Explosion Limit 1.6 - 7.7 %(V)
Density 0.81 g/cm3 (20 °C)
pH 6.2 (50 g/l, H2O, 20 °C)
Ignition Point 425 °C
Synonyms 3-Pentanone, Ethyl ketone
Hill Formula C5H10O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR