Ethylenediammonium dichloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ethylenediamine dihydrochloride

M-Clarity™ quality level = MQ200

8.00948.0005 8.00948.0250 8.00948.1000
EM8.00948.0005CS 30833 USD
EM8.00948.0005 EM8.00948.0250 EM8.00948.1000
Ethylenediammonium dichloride for synthesis, Sigma-Aldrich®
Ethylenediammonium dichloride
Formula: C₂H₁₀Cl₂N₂
MW: 133.02 g/mol
MDL Number: MFCD00012524
CAS Number: 333-18-6

Order Now

Specification Test Results

Solubility 300 g/l (20 °C)
Melting Point >300 °C
Molar Mass 133.02 g/mol
Bulk Density 650 kg/m3
pH 5 (100 g/l, H2O, 20 °C)
Synonyms 1,2-Diaminoethane dihydrochloride, Ethylenediamine dihydrochloride
Hill Formula C2H10Cl2N2
Chemical Formula H2NCH2CH2NH2 * 2HCl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR