Tungsten hexacarbonyl for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00888.0010 8.00888.0025
EM8.00888.0010CS 188645 USD
EM8.00888.0010 EM8.00888.0025
Tungsten hexacarbonyl for synthesis, Sigma-Aldrich®
Tungsten hexacarbonyl
Formula: C₆O₆W
MDL Number: MFCD00011462
CAS Number: 14040-11-0
UN: 3466
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 150 °C
Molar Mass 351.91 g/mol
Boiling Point 175 °C (1013 hPa)
Vapor Pressure 1.4 hPa (67 °C)
Density 2.65 g/cm3 (20 °C)
Synonyms Tungsten carbonyl
Hill Formula C6O6W
Chemical Formula W(CO)6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR