9,10-Anthraquinone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Anthracene-9,10-dione, Anthraquinone

M-Clarity™ quality level = MQ200

8.00465.0100 8.00465.0500
EM8.00465.0100CS 621 USD
EM8.00465.0100 EM8.00465.0500
9,10-Anthraquinone for synthesis, Sigma-Aldrich®
9,10-Anthraquinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
MDL Number: MFCD00001188
CAS Number: 84-65-1

Order Now

Specification Test Results

Solubility < - 0.001 g/l (20 °C)
Melting Point 282 - 285 °C
Molar Mass 208.22 g/mol
Bulk Density 600 kg/m3
Boiling Point 379 - 381 °C
Vapor Pressure 1.33 hPa (190 °C)
Flash Point 185 °C
Ignition Point 650 °C
Synonyms 9,10-Dihydro-9,10-anthracenedione
Hill Formula C14H8O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR