Butyric acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Butanoic acid

M-Clarity™ quality level = MQ200

8.00457.0100 8.00457.1000 8.00457.2500
EM8.00457.0100CS 543 USD
EM8.00457.0100 EM8.00457.1000 EM8.00457.2500
Butyric acid for synthesis, Sigma-Aldrich®
Butyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 163.5
Melting Pt: -3
Flash Pt: 67
MDL Number: MFCD00002814
CAS Number: 107-92-6
UN: 2820
ADR: 8 III
Merck Index: 13,01593

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -8 - -5 °C
Molar Mass 88.1 g/mol
Boiling Point 162 - 165 °C (1013 mbar)
Vapor Pressure 0.56 hPa (20 °C)
Flash Point 69 °C
Refractive Index 1.3991 (20 °C)
Explosion Limit 2 - 12.3 %(V)
Density 0.96 g/cm3 (20 °C)
pH 2.5 (100 g/l, H2O, 20 °C)
Ignition Point 425 °C
Hill Formula C4H8O2
Chemical Formula CH3CH2CH2COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR