3-Aminophenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3-Hydroxyaniline, m-Aminophenol

M-Clarity™ quality level = MQ200

8.00420.0005 8.00420.0250 8.00420.1000
EM8.00420.0005CS 51975 USD
EM8.00420.0005 EM8.00420.0250 EM8.00420.1000
3-Aminophenol for synthesis, Sigma-Aldrich®
3-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00007786
CAS Number: 591-27-5
UN: 2512
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 26 g/l (20 °C)
Melting Point 122 °C
Molar Mass 109.13 g/mol
Bulk Density 700 kg/m3
Boiling Point 164 °C (15 hPa)
Vapor Pressure < - 0.1 hPa (20 °C)
Flash Point 178 °C
Density 0.99 g/cm3 (25 °C)
pH 6.8 (10 g/l, H2O, 20 °C)
Ignition Point >600 °C
Synonyms 3-Hydroxyaniline
Hill Formula C6H7NO
Chemical Formula 3-(NH2)C6H4OH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR