2-Aminophenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Amino-1-hydroxy benzene, 2-Hydroxy aniline, 2-Hydroxyaniline, o-Aminophenol

M-Clarity™ quality level = MQ200

8.00419.0005 8.00419.0250 8.00419.1000 8.00419.9010
EM8.00419.0005CS 32468 USD
EM8.00419.0005 EM8.00419.0250 EM8.00419.1000 EM8.00419.9010
2-Aminophenol for synthesis, Sigma-Aldrich®
2-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00007690
CAS Number: 95-55-6
UN: 2512
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 17 g/l (20 °C)
Melting Point 172 - 174 °C (sublimed)
Molar Mass 109.12 g/mol
Bulk Density 600 kg/m3
Boiling Point 150 °C (15 hPa)
Vapor Pressure 14 hPa (153 °C)
Flash Point >175 °C
Density 1.328 g/cm3 (20 °C)
pH 6.5 - 7.5 (10 g/l, H2O, 20 °C)
Ignition Point 190 °C
Synonyms 2-Hydroxy aniline, 2-Amino-1-hydroxy benzene
Hill Formula C6H7NO
Chemical Formula 2-(NH2)C6H4OH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR