Myristic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Tetradecylic acid, Tetradecanoic acid

M-Clarity™ quality level = MQ200

8.00399.0100 8.00399.1000 8.00399.9050
EM8.00399.0100CS 960 USD
EM8.00399.0100 EM8.00399.1000 EM8.00399.9050
Myristic acid for synthesis, Sigma-Aldrich®
Myristic acid
Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
MDL Number: MFCD00002744
CAS Number: 544-63-8

Order Now

Specification Test Results

Solubility 1.07 mg/l (25 °C)
Melting Point 52 - 56 °C
Molar Mass 228.37 g/mol
Boiling Point 326 °C (1013 hPa)
Vapor Pressure < - 0.01 hPa (20 °C)
Flash Point >165 °C
Density 0.8622 g/cm3 (54 °C)
Synonyms Tetradecanoic acid
Hill Formula C14H28O2
Chemical Formula CH3(CH2)12COOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR