Ethyl octanoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate

M-Clarity™ quality level = MQ200

8.00202.0005 8.00202.0250 8.00202.9010
EM8.00202.0005CS 1850 USD
EM8.00202.0005 EM8.00202.0250 EM8.00202.9010
Ethyl octanoate for synthesis, Sigma-Aldrich®
Ethyl octanoate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Melting Pt: –47 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00009552
CAS Number: 106-32-1
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.070 g/l (25 °C)
Melting Point -47 °C
Molar Mass 172.27 g/mol
Boiling Point 208 °C (1013 hPa)
Vapor Pressure 0.2 hPa (20 °C)
Flash Point 79 °C
Refractive Index 1.4166 (20 °C, 589 nm)
Explosion Limit 0.7 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 325 °C
Synonyms Ethyl caprylate, Octanoic acid ethyl ester
Hill Formula C10H20O2
Chemical Formula CH3(CH2)6COOC2H5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR