Acrylic acid stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Prop-2-enoic acid

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.00181.0100 8.00181.0500 8.00181.1000
EM8.00181.0100CS 2381 USD
EM8.00181.0100 EM8.00181.0500 EM8.00181.1000
Acrylic acid stabilised for synthesis, Sigma-Aldrich®
Acrylic acid
Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 141.6
Melting Pt: 13
MDL Number: MFCD00004367
CAS Number: 79-10-7
UN: 2218
ADR: 8 (3) II

Order Now

Specification Test Results

Solubility 1000 g/l (25 °C)
Melting Point 13 °C
Molar Mass 72.06 g/mol
Boiling Point 141 °C (1013 hPa)
Vapor Pressure 3.8 hPa (20 °C)
Flash Point 46 °C
Refractive Index 1.4224 (20 °C, 589 nm)
Explosion Limit 3.9 - 19.8 %(V)
Density 1.05 g/cm3 (20 °C)
pH 2.1 (72.06 g/l, H2O, 20 °C)
Ignition Point 390 °C DIN 51794
Synonyms 2-Propenoic acid, Acroleic acid
Hill Formula C3H4O2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR