Dimethyl adipate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester

M-Clarity™ quality level = MQ200

8.00097.0250 8.00097.1000
EM8.00097.0250CS 541 USD
EM8.00097.0250 EM8.00097.1000
Dimethyl adipate for synthesis, Sigma-Aldrich®
Dimethyl adipate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 229
Melting Pt: 10.3
Flash Pt: 107
MDL Number: MFCD00008469
CAS Number: 627-93-0
Merck Index: 13,00163

Order Now

Specification Test Results

Solubility < - 1 g/l (20 °C)
Melting Point 8 - 11 °C
Molar Mass 174.19 g/mol
Boiling Point 215 - 225 °C (1013 hPa)
Vapor Pressure 0.06 hPa (20 °C)
Flash Point 107 °C
Refractive Index 1.4285 (20 °C, 589 nm)
Explosion Limit 0.81 - 8.1 %(V)
Density 1.06 g/cm3 (20 °C)
Ignition Point 360 °C
Viscosity kinematic 2.5 mm2/s (25 °C)
Synonyms Dimethyl hexanedioate, Adipic acid dimethyl ester
Hill Formula C8H14O4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR