Assay Diluent (Protein-Free), Surmodics IVD, Inc.
Supplier: Surmodics
SM01-0050
SM01-1000
102466-318EA
43.08
USD
102466-318
102466-320
Assay Diluent (Protein-Free), Surmodics IVD, Inc.
Buffers
Electrophoresis Buffers
Blocking Buffers
The Surmodics® Assay Diluent (Protein-Free) is a universal blocking agent that utilizes synthetic components to reduce matrix interference within immunoassay applications. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies (HAMA and rheumatoid factor).
- Protein-free
- Reduces non-specific binding
- Reduces matrix interfernece
- Increased signal-to-noise ratios
Surmodics protein-free assay diluent reduces matrix interference and provides accurate dilution recovery of patient specimens.
Certificaciones: ISO 13485:2016 & 9001:2015 Certified.