ActivArmr® 70-761 Cut resistant gloves, Ansell
Supplier: Ansell Healthcare
245709 245712 245708 245711
89067-982CS 1535.54 USD
89067-982 89067-988 89067-980 89067-986
ActivArmr® 70-761 Cut resistant gloves, Ansell
Gloves
Featuring revolutionary yarns for increased cut protection without the bulk, 70-761 Medium-Weight 10-gauge gloves are engineered with a cut protection yarn that provides the highest cut protection-to-weight ratio available.
  • Excellent flexibility
  • Superior cut protection
  • Maximum durability in high-wear conditions
Manufactured with DuPont™ Kevlar® fibers, gloves may be used in environments with intermittent heat contact up to 200 °F.
Use in the appliance manufacturing, automotive, general carpentry, general industry, HVAC/Sheet Metal Users/Window Glazers, metal fabrication, metalworking, paper, and plastics industries. Uses include metal finish; press, body shop; automated welding feeding; handling small and medium metal pieces with sharp edges; and assembly.
Gloves are designed to fit the natural contours of the hand, providing excellent flexibility for their category of cut protection (ANSI Cut Level 4). 100% Kevlar® reinforced thumb crotch delivers maximum durability in this high-wear area. Gloves are machine washable, which extends glove life.
Certificaciones: ANSI level 4 cut protection; ANSI level 2 abrasion protection.
Envase: Gloves are packaged 12 pairs in a bag; 6 bags in a carton.