LABODISC 50 PTFE Vent Filter, Advantec
Supplier: Advantec MFS
50JP020AN 50JP050AN
101103-570PK 127.89 USD
101103-570 101103-572 101104-852
LABODISC 50 PTFE Vent Filter, Advantec
Filters Venting Filters
Disposable, convenient 50 mm filter unit features a hydrophobic PTFE membrane in a high purity polypropylene housing.
  • Autoclavable
  • Act as an absolute retentive membrane
Use as a sterile air vent for fermentation tanks, bioreactors, or receiving containers. Protect pumps from aqueous liquids and aerosols.
Connections are 7 to 13.5 mm stepped hose barb with male luer slip. Housing has a vent port.
Información para pedidos: Sterile units (model 50JP020AS) are individually bagged.