Poly-D-lysine hydrobromide, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Poly-lysine is a polycation which binds to DNA, red cell membrane and any negatively charged protein. When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding. A compromise between the easier to use lower molecular weight products and the extremely viscous higher molecular weights would be the products in the range of 70,000-150,000.
It is typically used as a coating substrate for culture dishes, slides, etc. It enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. Both the D- and L- form of the poly-lysine can be used as a coating substrate since poly-lysine is a nonspecific attachment factor for cells; however, certain cells can digest poly-lysine. In this case, poly-D-lysine should be used as the attachment factor so that the cells are not disrupted by excessive uptake of L-lysine.
Other uses for poly-lysine have been reported as well:
• Conjugation to methotrexate for increased drug transport.
• Microencapsulation of islets.
• Use in simple reproducible procedure for chromosomal preparations from a variety of tissues.
• Immobilation of living cells in biocompatible semipermeable microcapsules.
• In the preparation of polycationic beads.
• Conjugation to albumin and horseradish peroxidase to enhance cellular uptake.
Poly-D-lysine hydrobromide is a synthetic amino acid that enhances cell adhesion to solid substrates. It has additionally been shown to eliminate prion proteins from infected cells.
- Presentation: White Powder
- Solubility: 50 mg/mL, H20
- Storage temperature: -20°C
Poly-lysine with molecular weight
Poly-D-lysine hydrobromide is a synthetic amino acid that enhances cell adhesion to solid substrates. It has additionally been shown to eliminate prion proteins from infected cells.
Formula:
C₆H₁₅BrN₂O₂ MW: 227.102 g/mol Storage Temperature: Freezer |
CAS Number:
27964-99-4 |
Specification Test Results
Identity Test | Passes |
Avg. Molecular Weight | 70,000-150,000 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...