Bio-Rad® Protein Assays, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
The Bio-Rad® Protein Assay is a simple colorimetric assay for measuring total protein concentration, based on the Bradford dye-binding method
This protein assay is adaptable to a range of protein concentrations and sample volumes. The standard procedure is used with protein concentrations between 0.2 and 1.4 mg/ml and 20–140 µg total protein. The microassay procedure is for protein concentrations of less than 25 µg/ml and 1–20 µg total protein. For rapid protein determination the assay can be carried out in 96-well microplates.
The kits provide the option of either bovine serum albumin or bovine γ-globulin standards.