S1000™ Thermal Cycler, Bio-Rad
Supplier: Bio-Rad
1840138 1840148 1840196 1840197
101645-580EA 6325 USD
101645-580 100069-188 100069-190 100069-192 100069-194 100446-338 100446-340 100446-342 100446-344
S1000™ Thermal Cycler, Bio-Rad
Thermal Cyclers
The flexible S1000™ Thermal Cycler has the choice of four interchangeable modules, and can be used as a stand-alone instrument for PCR or connected to a C1000 Touch™ Thermal Cycler for additional throughput
Reduced-mass honeycomb sample block delivers fast ramping and settling times. Fully adjustable heated lid offers complete sealing with a wide range of sized of reaction vessels. Programmable temperature gradient provides fast, easy determination of optimal annealing temperatures.
The four interchangeable reaction modules for the S1000™ Thermal Cycler include a gradient-enabled 96-well fast module, a gradient-enabled 96–deep well module, a gradient-enabled dual 48/48-well fast module that allows 2 independently controlled protocols to be run side by side in a single bay, and a gradient-enabled 384-well module.
The modules can be quickly swapped without tools. Up to three S1000™ Thermal Cyclers can be connected to a C1000 Touch™ Cycler, and controlled as one high-throughput instrument. You can also add PC control for up to 32 cyclers
Temperature control modes: Calculated and Block