Anti-CD8A Mouse Monoclonal Antibody (violetFluor® 450) [clone: SK1]
Supplier: Tonbo Biosciences
75-0087-T025 75-0087-T100 75-0087-T500
10208-486EA 122.1 USD
10208-486 10208-488 10208-490
Anti-CD8A Mouse Monoclonal Antibody (violetFluor® 450) [clone: SK1]
Antibodies
The SK1 antibody is specific for the 32-34 kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development,More Product Information