Cod liver oil, clear yellow liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0290140501 0290140504
IC90140501EA 83.53 USD
IC90140501 IC90140504
Cod liver oil, clear yellow liquid
Cod liver oil

Cod liver oil is a nutritional supplement derived from liver of cod fish, it has high levels of the omega-3 fatty acids, EPA and DHA.


  • Vitamin A = >850 USP u/gm
  • Vitamin D = >85 USP u/gm


Slightly soluble in alcohol; freely soluble in chloroform, ether, carbon disulphide, ethyl acetate and petrolium ether.

Storage Temperature: Ambient
MDL Number: MFCD00146493
CAS Number: 8001-69-2

Order Now

Specification Test Results

Form Clear yellow liquid
Storage Store at Room Temperature (15-30 °C)
Source Livers of cod


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR