Prostaglandin E2 ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219546101 0219546105
IC19546101EA 184.76 USD
IC19546101 IC19546105
Prostaglandin E2 ≥99%, powder
Prostaglandin E2

PGE2 is one of the primary cyclooxygenase products of arachidonic acid and one of the most widely investigated prostaglandins. Its activity influences inflammation, fertility and parturition, gastric mucosal integrity, and immune modulation. The effects of PGE2 are transduced by at least four distinct receptors designated EP1, EP2, EP3, and EP4. Affinity constants (Kd) of PGE2 for these receptors range from 1-10 nM depending on the receptor subtype and tissue.


Prostaglandin E2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation.


Sparingly soluble in water but freely soluble in organic solvents such as ethanol, methanol, acetone (clear solution), acetonitrile, or DMSO. The solubility of PGE2 in these solvents is at least 10 mg/mL. It is stable for at least 6 months in these solvents if stored at -20 °C.


Most biologically active prostaglandin. PGE2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation; regulates adenylyl cyclase. Tumor cells that over-express cyclooxygenase 2 display increased invasiveness, angiogenesis, and resistance to apoptosis that may be due to the PGE2-induced expression of angiogenic factors and stabilization of the anti-apoptotic protein, survivin.The effect of PGE2 on the immune system is mixed. It inhibits T cell activation in vitro, suggesting it is an immunosuppressant. However, in vivo, it appears to effect expansion of the Th17 subset and differentiation of the Th1 subset of T helper cells, marking it as an immunoactivator.

Formula: C₂₀H₃₂O₅
MDL Number: MFCD00077861
CAS Number: 363-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b537eb3c566d28c02d38413b3790362b51c38f80&vid=b537eb3c566d28c02d38413b3790362b51c38f80&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR