D-(+)-Proline ≥99%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

02195456.2 0219545601 0219545605
IC195456.2EA 58.34 USD
IC195456.2 IC19545601 IC19545605
D-(+)-Proline ≥99%, white powder
D-(+)-Proline

Soluble in water (5% - clear colorless to very faint yellow solution); slightly soluble in acetone, ethanol.


Soluble in water (5% - clear colorless to very faint yellow solution); slightly soluble in acetone, ethanol.

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 225…235 °C
MDL Number: MFCD00064317
CAS Number: 344-25-2

Order Now

Specification Test Results

Optical Rotation +84.5° to +86.5° (c=4, H2O)
Purity ≥99%
FTIR Conforms to standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR