Polyethylene glycol 8,000, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000

02195445.5 0219544501 0219544502
IC195445.5EA 84.33 USD
IC195445.5 IC19544501 IC19544502
Polyethylene glycol 8,000, powder
Polyethylene glycol 8,000

Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water.


PEG has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules, and in induction of cell hybridisation. PEG 8 000, can be used to precipitate DNA. PEG has also been used to recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples. PEG has demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis.


PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds. PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives


Soluble in aqueous solution (50 mg/ml - clear, colorless solution).


Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water. PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds. PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives.

Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250
MDL Number: MFCD01779615
CAS Number: 25322-68-3
Merck Index: 12,07729

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 56 - 66 °C
pH (5% aq soln) 5.0 - 7.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR