Glucose Oxidase (from Aspergillus niger), MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219519650 0219519610 0219519683 0219519690
IC19519650EA 139.16 USD
IC19519650 76177-448 76177-450 76177-452
Glucose Oxidase (from Aspergillus niger), MP Biomedicals
Enzymes

Glucose oxidase is an FAD-containing glycoprotein. The enzyme is specific for β-D-glucose. O can be replaced by hydrogen acceptors such as 2,6-dichlorophenol indophenol. Glucose oxidase from Aspergillus niger is a dimer consisting of 2 equal subunits with a molecular mass of 80 kDa each. Each subunit contains one flavin adenine dinulceotide moiety and one iron. The enzyme is a glycoprotein containing ~16% neutral sugar and 2% amino sugars. The enzyme also contains 3 cysteine residues and 8 potential sites for N-linked glycosylation. Glucose oxidase is capable of oxidizing D-aldohexoses, monodeoxy-D-glucoses, and methyl-D-glucoses at varying rates. Glucose oxidase does not require any activators, but it is inhibited by Ag+, Hg2+, Cu2+, phenylmercuric acetate, and p-chloromercuribenzoate. It is not inhibited by the nonmetallic SH reagents: N-ethylmaleimide, iodoacetate, and iodoacetamide.


  • Presentation: Yellow Lyophilized Powder
  • Isoelectric Point (pl): 4.2 (Lit.)
  • Extinction Coefficient (E1%): 16.7 (280 nm)(Lit.)
  • pH Optimum: 6.5, pH Stability: 8.0, pH Range: 4-7
  • Inhibitors: Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, D-arabinose (50%). FAD binding is inhibited by several nucleotides. Thermal Stability: Below 40 °C , Catalase Activity <10 u/mg
  • Dissolves readily at 5 mg/mL in 0.1 M potassium phosphate pH 7.0, giving a clear, yellow solution, also soluble in water.


Glucose oxidase can be utilized in the enzymatic determination of D-glucose in solution. As glucose oxidase oxidizes β-D-glucose to D-gluconolactate and hydrogen peroxide, horseradish peroxidase is often used as the coupling enzyme for glucose determination. Although glucose oxidase is specific for β-D-glucose, solutions of D-glucose can be quantified as α-D-glucose will mutorotate to β-D-glucose as the β-D-glucose is consumed by the enzymatic reaction. Glucose oxidase is widely used in the food and pharmaceutical industries as well as a major component of glucose biosensors.


Glucose oxidase catalyses the oxidation of β-d-glucose to d-glucono-β-lactone and hydrogen peroxide, with molecular oxygen as an electron acceptor.


Inhibitors: Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, D-arabinose (50%). FAD binding is inhibited by several nucleotides. Thermal Stability: Below 40 °C , Catalase Activity \<10 u/mg


Glucose oxidase is an FAD-containing glycoprotein. The enzyme is specific for β-D-glucose. O can be replaced by hydrogen acceptors such as 2,6-dichlorophenol indophenol.Glucose oxidase from Aspergillus niger is a dimer consisting of 2 equal subunits with a molecular mass of 80 kDa each. Each subunit contains one flavin adenine dinulceotide moiety and one iron. The enzyme is a glycoprotein containing ~16% neutral sugar and 2% amino sugars. The enzyme also contains 3 cysteine residues and 8 potential sites for N-linked glycosylation. Glucose oxidase is capable of oxidizing D-aldohexoses, monodeoxy-D-glucoses, and methyl-D-glucoses at varying rates. Glucose oxidase does not require any activators, but it is inhibited by Ag+, Hg2+, Cu2+, phenylmercuric acetate, and p-chloromercuribenzoate. It is not inhibited by the nonmetallic SH reagents: N-ethylmaleimide, iodoacetate, and iodoacetamide.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ab54a179e57bca6a87f04a8b65c48f3ee407f2c&vid=d57a2780f87ed1b090ed366c14d4e57680dd3de7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d4b4850b64e56d298ebd5b99709eb25dc4ce049&vid=c0e1d9b11edbb0f4821d37cac9996a2ecd698b2b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Enzyme Name Glucose oxidase
Source Aspergillus niger
CAS No. 9001-37-0
Form Yellow lyophilized powder
Storage Conditions Store at -20 °C, desiccated
Enzyme Activity ≥100 u/mg material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR