Fluorescamine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219518325 0219518380 0219518391 0219518383
IC19518325EA 62 USD
IC19518325 IC19518380 IC19518391 76177-446
Fluorescamine, white powder
Fluorescamine

Fluorescamine is a non-fluorescent reagent that reacts readily with primary amino groups in amino acids and peptides to form highly fluorescent derivatives, whereas the reagent and its degradation products are non-fluorescent. No significant fluorescence with ammonia. Low background due to hydrolysis.
an plasma and urine and sulfonamides in honey by HPLC with fluorescence detection.


  • Fluorescence: Emmax: 475 nm; Exc.: 390 nm in 0.5 M borate buffer, pH 8.5, after derivatization with L-leucine


Fluorescamine is a reagent for fluorescent assay of amino acids, peptides, proteins and other primary amines. It is also used for fluorometric assays of some proteolytic enzymes. It effectively blocks newly generated amino termini in protein sequence analyses by the Beckman automated sequencer. This product has been used in labeling casein so that it can be used as a substrate for measuring protease activity.


Fluorescamine is used in many sensitive detection methods, eg. characterization of poly-L-lysine (pLL)/DNA complexes post-modified with a multivalent hydrophilic polymer, spectrofluorimetric analysis of procaine, detection and quantitation of residual aminopenicillins by HPLC after fluorescamine derivation, determination of lisinopril in hum


Soluble in CH₂Cl₂, trichloromethane, chloroform, dimethysulfoxide; slightly soluble in acetone (50 mg/ml), ethanol and water.


Fluorescence: Emmax: 475 nm; Exc.: 390 nm in 0.5 M borate buffer, pH 8.5, after derivatization with L-leucine.

Formula: C₁₇H₁₀O₄
Melting Pt: 148…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00005928
CAS Number: 38183-12-9

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR