n-Dodecane ≥99%, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
0219515880
0219515890
0219515891
0219515825
IC19515880EA
139.62
USD
IC19515880
IC19515890
IC19515891
76177-442
n-Dodecane ≥99%, liquid
n-Dodecane
N-Dodecane is a liquid alkane hydrocarbon.
N-Dodecane is used as a solvent, distillation chaser, scintillator component. Moreover it is used as a diluent for tributyl phosphate (TBP) in plants reprocessing.
- Presentation: Clear, Colorless Liquid
- Auto Ignition: 203 °C (397 °F) (lit.)
- Vapor Density: 5.96 (vs air) (Lit.)
- Vapor Pressure: 1 mmHg ( 47.8 °C) (Lit.)
- Refractive Index: n20/D 1.421(lit.)
- Storage temperature: Store at Room Temperature (15-30 °C). Store Desiccated.
Formula:
H₃C(CH₂)₁₀CH₃ Boiling Pt: 215…217 °C (lit.) Melting Pt: –9.6 °C (lit.) Density: 0.753 (25 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00008969 CAS Number: 112-40-3 UN: 1993 ADR: 3,III |
|
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...