1,3-Dielaidin ≥99%, liquid
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219515180
0219515190
IC19515180EA
149.49
USD
IC19515180
IC19515190
1,3-Dielaidin ≥99%, liquid
1,3-Dielaidin
Formula:
C₃₉H₇₂O₅ |
MDL Number:
MFCD00042913 CAS Number: 25637-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...