Cetrimonium bromide, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

0219502980 0219502990 0219502991 0219502905
IC19502980EA 89.21 USD
IC19502980 IC19502990 IC19502991 IC0219502905
Cetrimonium bromide, white powder
Cetrimonium bromide

A phase transfer catalyst ideal for the precipitation of DNA


Cetyltrimethylammonium bromide ia used as an cationic surfactant suitable for the isolation of high molecular weight DNA in plants and other organisms(by a rapid method) as well as plant DNA for use in PCR analysis. It has been used to precipitate nucleic acids. CTAB provides enhancement of Concanavalin A mediated agglutination. It has been used for the determination of protein molecular weights in electrophoretic systems and for determination of critical micelle concentration of detergents. CTAB has also been used as a titrant for potentiometric titration of perchlorate and as a phase-transfer catalyst in reduction of arenes and heterocyclic compounds.


Freely but slowly soluble in water; Freely soluble in methanol and ethanol; soluble in alcohol.


Cetyltrimethylammonium bromide ia used as an cationic surfactant suitable for the isolation of high molecular weight DNA in plants and other organisms(by a rapid method) as well as plant DNA for use in PCR analysis. It has been used to precipitate nucleic acids. CTAB provides enhancement of Concanavalin A mediated agglutination. It has been used for the determination of protein molecular weights in electrophoretic systems and for determination of critical micelle concentration of detergents. CTAB has also been used as a titrant for potentiometric titration of perchlorate and as a phase-transfer catalyst in reduction of arenes and heterocyclic compounds.


Precaución: Research Use Only (RUO).

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Flash Pt: 244
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
UN: 3077
ADR: 9 III
Merck Index: 13,02034
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d254936cbb112618d1f5c2073ee43f98e2bd2fc8&vid=d254936cbb112618d1f5c2073ee43f98e2bd2fc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes Passes
FTIR Conforms to Standard
Storage Store at Room Temperature (15-30 °C).
Aggregation Number 170
CMC 1 mM (20-25 °C)
pH 6.0-7.5 (0.1 M H2O, 0 °C)(Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR