Ricinoleic acid ≥99%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid

0219151480 0219151490 0219151491
IC19151480EA 105.82 USD
IC19151480 IC19151490 IC19151491
Ricinoleic acid ≥99%, liquid
Ricinoleic acid

Ricinoleic Acid is a naturally occurring 12-hydroxy fatty acid constituting about 90% of the fatty acids in castor oil. Has the capacity to serve as a substrate for the synthesis of the conjugated linoleic acids.


Ricinoleic Acid is used as a substrate for the synthesis of the conjugated linoleic acids.


Ricinoleic Acid is a naturally occurring 12-hydroxy fatty acid constituting about 90% of the fatty acids in castor oil. Has the capacity to serve as a substrate for the synthesis of the conjugated linoleic acids.

Formula: C₁₈H₃₄O₃
MDL Number: MFCD00084840
CAS Number: 141-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR