D-(-)-N-Methylglucamine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol

0219150690 0219150691
IC19150690EA 172.13 USD
IC19150690 IC19150691
D-(-)-N-Methylglucamine, white powder
D-(-)-N-Methylglucamine

N-Methylglucamine is used in the synthesis of surface active agents, pharmaceuticals, dyes; an antimonate as antiprotozoal. Used as a Na+ substitute in systems to maintain physiological solutions as isotonic but without sodium. Has also been used in the studies of rabbit cerebral artery, rabbit cardiac myocytes, perfused rat intestine, and frog skeletal muscle.


Soluble in water (50 mg/ml - clear, colorless solution).


Used in the synthesis of surface active agents, pharmaceuticals, dyes; an antimonate as antiprotozoal. Used as a Na+ substitute in systems to maintain physiological solutions as isotonic but without sodium. Has also been used in the studies of rabbit cerebral artery, rabbit cardiac myocytes, perfused rat intestine, and frog skeletal muscle.

Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
MW: 195.22 g/mol
Melting Pt: 125…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00004707
CAS Number: 6284-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR