Folinic acid calcium salt pentahydrate, pale yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Calcium folinate pentahydrate, Calcium (4-(((2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzoyl)-L-glutamate pentahydrate, Calcium (2S)-2-(4-(((2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioate pentahydrate

0219150480 0219150491
IC19150480EA 175.9 USD
IC19150480 IC19150491
Folinic acid calcium salt pentahydrate, pale yellow powder
Folinic acid calcium salt pentahydrate

Folinic acid calcium salt, pentahydrate offsets the effects of folic acid antagonists that act by binding dihydrofolate reductase and blocking the conversion of folic acid to tetrahydrofolate.


In medicine, calcium folinate is used after treatment with methotrexate to reduce the toxicity of the methotrexate. Calcium folinate is also used to enhance the cytotoxic effect of 5-fluorouracil in the treatment of cancer.


Intermediate product of the metabolism of folic acid; the active form into which that acid is converted in the body, ascorbic acid being a necessary factor in the conversion process.


Freely soluble in water (100 mg/mL-clear to slightly hazy solution); almost insoluble in ethanol, alcohol.

Formula: C₂₀H₃₁CaN₇O₁₂
MW: 601.583 g/mol
MDL Number: MFCD00149465
CAS Number: 6035-45-6

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR