Cellulose 97.0-102.0%, microcrystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219149980 0219149983 0219149991
IC19149980EA 51.75 USD
IC19149980 IC19149991 IC19149983
Cellulose 97.0-102.0%, microcrystalline powder
Cellulose

Cellulose is an organic compound, a polysaccharide consisting of a linear chain of several hundred to over ten thousand β(1→4) linked D-glucose units.


Cellulose is used as bulking agent, opacifier, anti-caking agent, extrusion aid and stabiliser for foams and emulsions. High purity cellulose powders are used for partition chromatography.


Soluble in ether, insoluble in water (Water soluble substances <12,0 mg/5 g)

Formula: (C₁₂H₂₂O₁₁)n
MW: 342.3 g/mol
MDL Number: MFCD00081512
CAS Number: 9004-34-6

Order Now

Specification Test Results

Identity Test Passes Test
Purity 97.0 - 102.0%
Moisture content 5.0%
Particle Size +60 mesh (wt%) 8%
Particle Size +200 mesh (wt%) 45%
pH 5 - 7
Residue on ignition 0.05%
Water Soluble Substances 12.0 mg/5 g
Heavy Metals 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR