Zinc sulfate heptahydrate 99.0-103.0%, crystals ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
0219145291
0219145290
0219145280
IC19145291EA
141.48
USD
IC19145291
IC19145280
IC19145290
Zinc sulfate heptahydrate 99.0-103.0%, crystals ACS
Zinc sulphate heptahydrate
Formula:
ZnSO₄·7H₂O MW: 287.54 g/mol |
MDL Number:
MFCD00149894 CAS Number: 7446-20-0 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99,0 - 103,0% |
pH (5% aq soln) | 4.4 - 6.0 |
Insoluble Matter | ≤0.01% |
Chloride | ≤5 ppm |
Nitrate | ≤0.002% |
Ammonium | ≤0.001% |
Calcium | ≤0.005% |
Iron | ≤0.001% |
Lead | ≤0,003% |
Magnesium | ≤0.005% |
Manganese | ≤3 ppm |
Potassium | ≤0.01% |
Sodium | ≤0.05% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...