Sodium dihydrogen phosphate monohydrate 98,0-102,0, powder_white ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
02191442.1
02191442.5
0219144201
0219144205
IC191442.1EA
65.05
USD
IC191442.1
IC191442.5
IC19144201
IC19144205
Sodium dihydrogen phosphate monohydrate 98,0-102,0, powder_white ACS
Sodium dihydrogen phosphate monohydrate
Sodium phosphate monobasic monohydrate is useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Freely soluble in water; insoluble in ethanol.
Precaución: Research Use Only (RUO).
Formula:
NaH₂PO₄·H₂O MW: 137.99 g/mol Melting Pt: 100 |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 Merck Index: 13,08734 |
|
Specification Test Results
Identity Test | Passes |
Purity | 98.0% - 102.0% |
pH | 4,1 - 4,5 (5% aq soln) |
Insoluble Matter | ≤0,01% |
Chloride | ≤5 ppm |
Sulfate | ≤0,003% |
Calcium | ≤0,005% |
Potassium | ≤0,01% |
Heavy Metals | ≤0,001% |
Iron | ≤0,001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...