Disodium hydrogen phosphate heptahydrate ≥98%, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

02191441.5 0219144101 0219144105
IC191441.5EA 110.92 USD
IC191441.5 IC19144101 IC19144105
Disodium hydrogen phosphate heptahydrate ≥98%, white crystalline powder ACS
Disodium hydrogen phosphate heptahydrate

Sodium phosphate dibasic heptahydrate is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Sodium phosphate dibasic is highly hygroscopic and water soluble. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.


Sodium phosphate dibasic is highly hygroscopic and water soluble. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers.


Sodium phosphate dibasic heptahydrate is use in many applications such as sequestrant, emulsifier and buffer in foods. As mordant in dyeing for weighting silk, in tanning. In manufacturing of enamels, ceramics, detergents; as fireproofing agent in soldering and brazing instead of borax; as reagent and buffer in analytical chemistry, cathartic, laxative.


Soluble in water, but more soluble in hot water; insoluble in alcohol, ethanol.


Chloride= ≤0.001%; Sulfate (SO4)= ≤0.005%; Iron (Fe)= ≤0.001%

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6

Order Now

Specification Test Results

Purity ≥98%
pH (5% aq soln) 8.7 - 9.3
Insoluble Matter ≤0,005%
Heavy Metals ≤0,001%
Chloride ≤0,001%
Sulphate (SO4) ≤0,005%
Iron (Fe) ≤0,001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR