di-Sodium tetraborate decahydrate 99.5-105.0%, white crystalline powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
Sodium borate decahydrate is a mineral, and a salt of boric acid. The term borax is used for a number of closely related minerals or chemical compounds that differ in their crystal water content, but usually refers to the decahydrate.
Applications: Sodium borate is used in biochemical and chemical laboratories to make buffers, e.g. for gel electrophoresis of DNA, such as TBE(Tris/Borate/EDTA) or SB(Sodium borate) buffer or BBS (borate buffered saline) in coating procedures. As a micronutrient fertilizer to correct boron-deficient soils. To clean the brain cavity of a skull for mounting.
1 g dissolves in 16 M water, 0.6 mL boiling water, 1 mL glyceral; insoluble in alcohol.
Formula:
Na₂B₄O₇·10H₂O MW: 381.37 g/mol Boiling Pt: 320 Melting Pt: 75 |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 Merck Index: 13,08662 |
Specification Test Results
Assay | 99.5 - 105.0% |
Heavy Metals | ≤0.001%(as Pb) |
Insoluble Matter | ≤0.005% |
pH | 9.15 - 9.20 (0.01M aq soln) |
Chloride (Cl) | ≤0.001% |
Sulphate (SO4) | ≤0.005% |
Phosphate (PO4) | ≤0.001% |
Calcium (Ca) | ≤0.005% |
Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...