Dipotassium hydrogen orthophosphate trihydrate, white crystalline powder, Reagent Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate
Potassium phosphate is a reagent with high buffering capacity. It occurs in several forms: monobasic, dibasic and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.
- Component of Culture media
- Component in phosphate buffered saline
Potassium phosphate dibasic is widely used in molecular biology, biochemistry, and chromatography. It is typically used as a component for a wide variety of media used in the culture of microorganisms, also as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.
Soluble in water(100 mg/ml); Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Precaución: Research Use Only (RUO).
Formula:
K₂HPO₄·3H₂O MW: 228.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149926 CAS Number: 16788-57-1 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Reference |
pH (5% aq soln) | 8.5 - 9.6 |
Solubility (0.2 g/ml in water) | Clear, colorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...