Potassium acetate ≥99.0%, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Acetic acid potassium salt

0219142580 0219142590 0219142591
IC19142580EA 56.78 USD
IC19142580 IC19142590 IC19142591
Potassium acetate ≥99.0%, white crystalline powder ACS
Potassium acetate

Potassium acetate is utilized in the isolation of RNA intermediates and products from in vitro splicing reactions performed with a HeLa cell nuclear extract.Potassium acetate is used as a buffer in molecular biology research, notably in the isolation of DNA.


Solubility: H₂O: 1 M at 20 °C, clear, colorless

Formula: CH₃COOK
MW: 98.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥99,0%
Heavy Metals <5 ppm
Insoluble Matter <0,005%
pH 6,5 - 9,0 (5% aq soln)
Chloride <0,003%
Phosphate <0,001%
Sulphate <0,002%
Iron <5 ppm
Calcium <0,005%
Magnesium <0,002%
Sodium <0,03%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR