Benzoic acid ≥99.5%, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0219141280
0219141290
0219141291
IC19141280EA
96.74
USD
IC19141280
IC19141290
IC19141291
Benzoic acid ≥99.5%, powder ACS
Benzoic acid
Insoluble in methanol.
Freezing Point 122 - 123 °C; Insoluble in Methanol ≤0.005%; Chlorine Compounds ≤0.005%; Sulfur Compounds ≤0.002%
Formula:
C₇H₆O₂ MW: 122.12 g/mol |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 |
Specification Test Results
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). |
Heavy Metals | ≤5 ppm |
Residue on Ignition | ≤0.005% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...