Barium chloride dihydrate ≥99.0%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Barium dichloride

0219141080 0219141090
IC19141080EA 67.37 USD
IC19141080 IC19141090
Barium chloride dihydrate ≥99.0%, powder ACS
Barium chloride dihydrate

Soluble in water.


Oxidizing Substances (as NO3) ≤0.005%, Calcium ≤0.05%, Potassium ≤0.0025%, Sodium ≤0.005%, Strontium ≤0.1%, Iron ≤2 ppm

Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
MDL Number: MFCD00149154
CAS Number: 10326-27-9
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Form White Granular Powder
Storage Store at Room Temperature (15-30 °C).
Assay ≥99.0%
Heavy Metals ≤5 ppm
Insoluble Matter ≤0.005%
Loss on Drying 14.0 - 16.0%
pH 5.2 - 8.2(5% aq soln)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR