di-Ammonium oxalate monohydrate ≥99.0%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

0219140880 0219140890 0219140891
IC19140880EA 65.44 USD
IC19140880 IC19140890 IC19140891
di-Ammonium oxalate monohydrate ≥99.0%, powder ACS
di-Ammonium oxalate monohydrate

Chloride ≤0.002%; Iron ≤2 ppm; Sulfate ≤0.002%

Formula: (NH₄)₂C₂O₄·H₂O
MW: 142.11 g/mol
MDL Number: MFCD00149694
CAS Number: 6009-70-7
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Synonyms Ethanedioic Acid Monohydrate; Oxalic acid diammonium salt
Form White Powder
Storage Store at Room Temperature (15-30 °C).
Heavy Metals ≤5 ppm
Insoluble Matter ≤0.005%
Residue on Ignition ≤0.02%
Chloride ≤0.002%
Iron ≤2 ppm
Sulfate ≤0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR