Aluminium sulfate hydrate 98.0-102.0%, white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Aluminum(III) sulphate xhydrate

0219140380 0219140390 0219140391
IC19140380EA 48.96 USD
IC19140380 IC19140390 IC19140391
Aluminium sulfate hydrate 98.0-102.0%, white powder ACS
Aluminium sulphate hydrate

Chloride Content ≤0.005%; Calcium ≤0.01%; Magnesium ≤0.002%; Potassium ≤0.005%; Sodium ≤0.02% ; Iron (Fe) ≤0.002%

Formula: Al₂(SO₄)₃·xH₂O
MW: 366.21 g/mol
Density: 1.69 g/mL (25 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149136
CAS Number: 17927-65-0
EINECS: 233-135-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98.0 - 102.0%
Insoluble Matter <0.01%
Chloride Content <0.005%
Calcium <0.01%
Magnesium <0.002%
Potassium <0.005%
Sodium <0.02%
Heavy Metals (as Pb) <0.001%
Iron (Fe) <0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR