Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥90%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt
0219123083
0219123090
0219123091
IC19123083EA
96.62
USD
IC19123083
IC19123090
IC19123091
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥90%, white powder
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Energy-rich precursor in the enzymatic biosynthesis of RNA. A potent vasodilator. Induces contractile responses in some tissues.
Soluble in water (50 mg/ml - clear colourless solution).
Formula:
C₉H₁₂N₂Na₃O₁₅P₃ MW: 550.09 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00044310 CAS Number: 19817-92-6 EINECS: 243-347-5 |
Specification Test Results
Identity Test | Passes |
Purity | ≥90% |
OD250/OD260 | 0.75 ± 0.05 |
OD280/OD260 | 0.40 ± 0.05 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...