Vinblastine sulfate, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219028701 0219028705 0219028725
IC19028701EA 42.79 USD
IC19028701 IC19028705 IC19028725
Vinblastine sulfate, white powder
Vinblastine sulfate

Vinblastine Sulfate is an anti-tumor alkaloid.


  • An anti-tumor alkaloid
  • Depolymerises microtubules.
  • Used in cancer research


Plant alkaloid that inhibits microtubule assembly by binding tubulin and inducing self-association in spiral aggregates in a reaction that appears to be regulated by the C-terminus of β-tubulin and is enhanced by GDP and GTP.


Arrests the cell cycle in G2/M-phase by blocking mitotic spindle formation. Triggers Raf-1 activation, phosphorylation of bcl-2-family proteins, induction of p53 expression, and apoptosis in several tumor cell lines. Substrate of Pgp and CYP3A4.


Very slightly soluble in ethanol. Practically insoluble in ether. One part is sol in 10 parts of water, 50 parts of chloroform.


Vinblastine Sulfate is an anti-tumor alkaloid.

Formula: C₄₆H₆₀N₄O₁₃S
MDL Number: MFCD00082457
CAS Number: 143-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR