Retinoic acid all trans, yellow powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin

02190269.5 02190269.6 0219026901
IC190269.5EA 152.7 USD
IC190269.5 IC190269.6 IC19026901
Retinoic acid all trans, yellow powder
Retinoic acid

Retinoic acid (RA) is a vitamin A-derived, non-peptidic, small lipophilic molecule that mediates the functions of vitamin A required for growth and development. It affects gene transcription and modulates a wide variety of biological processes like Cell Proliferation, Differentiation, including Apoptosis. Retinoic Acid mediated gene transcription depends on the rate of transport of Retinoic Acid to target cells and the timing of exposure of Retinoic Acid to RARs (Retinoic Acid Receptors) in the target tissues


Retinoic Acid has been used in various differentiation protocols, including B-cells, T cells and neurons and applied clinically to treat cancer as a form of differentiation-induction therapy.


All -trans -Retinoic acid (ATRA) is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR). The bound RAR and RXR act as transcription factors that regulate the growth and differentiation of both normal and malignant cells. Cytochromes P450 (CYPs) catalyse the 4-hydroxylation of ATRA. Retinoic acid primes embryonic stem cells to become neurons.


Soluble in methanol, chloroform (40 mg/ml) or acetone (30 mg/ml), slightly soluble in ethanol; Insoluble in water. It is recommended that solutions be made fresh for each use.


Retinoic Acid has been used in various differentiation protocols, including B-cells, T cells and neurons and applied clinically to treat cancer as a form of differentiation-induction therapy.

Formula: C₂₀H₂₈O₂
MW: 300.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00001551
CAS Number: 302-79-4

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 173 - 183°C
l max (isopropanol + sulfuric acid) 353 ± 3 nm
Loss on Drying ≤0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR