Procaine hydrochloride, solid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219026080 0219026083
IC19026080EA 72.35 USD
IC19026080 IC19026083
Procaine hydrochloride, solid
Procaine hydrochloride

Soluble in water (clear, 50 mg/mL).

Formula: C₁₃H₂₁ClN₂O₂
MDL Number: MFCD00013000
CAS Number: 51-05-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8202bb12cab03cc6f9a3771dac05bca8cf936753&vid=8202bb12cab03cc6f9a3771dac05bca8cf936753&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR