2-(4-Isobutylphenyl)propanoic acid, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: (±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen

0219021601 0219021605 0219021610
IC19021601EA 72.54 USD
IC19021601 IC19021605 IC19021610
2-(4-Isobutylphenyl)propanoic acid, powder
2-(4-Isobutylphenyl)propanoic acid

Ibuprofen an anti-inflammatory and analgesic.


Ibuprofen is clinically useful as a nonsteroidal anti-inflammatory drug.


Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2. Also inhibits cyclooxygenase, PGH synthase-1 and PGH synthase-2. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23.


Freely soluble in acetone; dichloromethane, ethanol; soluble in methanol (at least 50 mg/mL-clear, colorless solution) and most organic solvents; Insoluble in water.


Ibuprofen an anti-inflammatory and analgesic.


Precaución: Research Use Only (RUO).

Formula: C₁₃H₁₈O₂
MW: 206.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010393
CAS Number: 15687-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=392703f61d4457957d53d31ae4556d8e4d1700c6&vid=392703f61d4457957d53d31ae4556d8e4d1700c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 72 - 82 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR