Diazolidinyl urea, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea

0219019125 0219019183
IC19019125EA 65.1 USD
IC19019125 IC19019183
Diazolidinyl urea, powder
Diazolidinyl urea

Diazolidinyl Urea is a mixture of different formaldehyde addition products including polymers. It is chemically related to imidazolidinyl urea.


Diazolidinyl Urea is used as an antimicrobial preservative in cosmetics. It acts as a formaldehyde releaser.


Soluble in water (50% w/v).


Diazolidinyl Urea is a mixture of different formaldehyde addition products including polymers. It is chemically related to imidazolidinyl urea.

Formula: C₈H₁₄N₄O₇
MW: 278.22 g/mol
MDL Number: MFCD03547942
CAS Number: 78491-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0da82688a19e1d8142fe82975036b1bb951ee64&vid=b0da82688a19e1d8142fe82975036b1bb951ee64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form White Powder
Storage Store at +4 °C.
pH 6.0 - 7.5 (1% aq soln)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR