Clomiphene citrate salt, off-white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219017401 0219017405 0219017410 0219017425
IC19017401EA 100.83 USD
IC19017401 IC19017405 IC19017410 IC19017425
Clomiphene citrate salt, off-white powder
Clomiphene citrate salt

Clomiphene Citrate Salt is a mixture of cis- and trans- isomers. Clomiphene citrate is a selective estrogen receptor modulator.


Freely soluble in methanol; slightly soluble in water and chloroform; sparingly soluble in ethanol (96%); practically insoluble in ether.


Clomiphene Citrate Salt is a mixture of cis- and trans- isomers. Clomiphene citrate is a selective estrogen receptor modulator.

Formula: C₃₂H₃₆ClNO₈
Melting Pt: 116.5 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00058322
CAS Number: 50-41-9

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR