Adenosine-5'-diphosphate monosodium salt (from equine muscle) ≥97%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Adenosine 5'-diphosphate sodium salt, xSodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate, ADP-Na, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate

02190149.1 02190149.5 0219014901
IC190149.1EA 70.73 USD
IC190149.1 IC190149.5 IC19014901
Adenosine-5'-diphosphate monosodium salt (from equine muscle) ≥97%, powder
Adenosine-5'-diphosphate monosodium salt
Formula: C₁₀H₁₄N₅NaO₁₀P₂
MW: 449.184 g/mol
MDL Number: MFCD00065987
CAS Number: 20398-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR